Diflunisal

Lieferant: Thermo Scientific
Achtung

Synonyme: 2',4'-Difluoro-4-hydroxy[1,1'-biphenyl]-3-carboxylic acid, 2',4'-Difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid

460890010 460890050
ACRO460890010EA 36.6 EUR
ACRO460890010 ACRO460890050
Diflunisal
Diflunisal
Formel: C₁₃H₈F₂O₃
Molecular Weight: 250,1976 g/mol
Schmelzpunkt: 207…209 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00057834
CAS-Nummer: 22494-42-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (60°C, 4 H) (vacuum)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR