Sodium diatrizoate 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Diatrizoate sodium, 3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid sodium salt, 3,5-Diacetamido-2,4,6-triiodobenzoic acid sodium salt, Diatrizoic acid sodium salt, Amidotrizoate sodium salt, Sodium 3,5-diacetamido-2,4,6-triiodobenzoate

460960050 460960250 460961000
ACRO460960050EA 42.4 EUR
ACRO460960050 ACRO460960250 ACRO460961000
Sodium diatrizoate 99%
Sodium diatrizoate
Formel: C₁₁H₈I₃N₂NaO₄
Molecular Weight: 635,898 g/mol
Schmelzpunkt: 261…262 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051544
CAS-Nummer: 737-31-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤7 %
Titration with HClO4 ≥98.5 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR