Chlorhexidin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide]
461020010
461020050
ACRO461020010EA
92.1
EUR
ACRO461020010
ACRO461020050
Chlorhexidin 98%
Chlorhexidin
Formel:
C₂₂H₃₀Cl₂N₁₀ Molecular Weight: 505,4466 g/mol Schmelzpunkt: 134…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009673 CAS-Nummer: 55-56-1 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 130 to 136 °C |
Water | ≤1 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...