Meglumine antimoniate

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Methyl-D-glucamine antimoniate

461140250 461141000
ACRO461140250EA 96.9 EUR
ACRO461140250 ACRO461141000
Meglumine antimoniate
Meglumine antimoniate
Formel: C₇H₁₇NO₅·HO₃Sb
Molecular Weight: 365,97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20922853
CAS-Nummer: 133-51-7
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Assay 30.0 to 34.0 % (Sb)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR