Nevirapin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 11-Cyclopropyl-4-methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one

461150010 461152500
ACRO461150010EA 183 EUR
ACRO461150010 ACRO461152500
Nevirapin 98%
Nevirapin
Formel: C₁₅H₁₄N₄O
Molecular Weight: 266.30 g/mol
Schmelzpunkt: 247…249 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00866928
CAS-Nummer: 129618-40-2

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Powder
Infrared spectrum Conforms
Melting point 244 to 248 °C
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR