Ondansetron hydrochloride dihydrate 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 9-Methyl-3-((2-methyl-1H-imidazol-1-yl)methyl)-1,2,3,9-tetrahydro-4H-carbazol-4-one hydrochloride dihydrate, GR 3832 HCl 2H2O, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one Hydrochloride, SN-37 HCl 2H2O, NSC665799 HCl 2H2O

461160010 461160050
ACRO461160010EA 58.2 EUR
ACRO461160010 ACRO461160050
Ondansetron hydrochloride dihydrate 98%
Ondansetron hydrochloride dihydrate
Formel: C₁₈H₁₉N₃O·HCl·₂H₂O
Molecular Weight: 365.86 g/mol
Schmelzpunkt: 178 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00374371
CAS-Nummer: 103639-04-9
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 176 to 180 °C
Water ≤10.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR