Oseltamivir phosphate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3R,4R,5S)-Ethyl 4-acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate phosphate

461170010 461170050
ACRO461170010EA 60.3 EUR
ACRO461170010 ACRO461170050
Oseltamivir phosphate 98%
Oseltamivir phosphate
Formel: C₁₆H₂₈N₂O₄·H₃PO₄
Molecular Weight: 410,40 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08059548
CAS-Nummer: 204255-11-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤2 %
HPLC ≥97.5 %
Specific optical rotation -27° to -35° (20°C, 589 nm) (1 % in water) (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR