Pyridostigmine bromide 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-((Dimethylcarbamoyl)oxy)-1-methylpyridin-1-ium bromide, 3-(Dimethylcarbamoyloxy)-1-methylpyridinium Bromide
461190010
461190050
461190250
ACRO461190010EA
56.1
EUR
ACRO461190010
ACRO461190050
ACRO461190250
Pyridostigmine bromide 98%
Pyridostigmine bromide
Formel:
C₉H₁₃BrN₂O₂ Molecular Weight: 261,12 g/mol Schmelzpunkt: 154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079283 CAS-Nummer: 101-26-8 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 153 to 157 °C |
Loss on drying | ≤2 % (1 g, 105 °C) |
Titration with HClO4 | 97.5 to 102.5 % (on dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...