Ritonavir 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: A-84538, ABT-538, Abbott 84538

461220010 461222500
ACRO461220010EA 360 EUR
ACRO461220010 ACRO461222500
Ritonavir 98%
Ritonavir
Formel: C₃₇H₄₈N₆O₅S₂
Molecular Weight: 720,94 g/mol
Schmelzpunkt: 120…132 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00927142
CAS-Nummer: 155213-67-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 121 to 127 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR