Ifosfamide
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Ifex, 3-(2-Chloroethyl)-2-((2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide, N,3-Bis(2-chloroethyl)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine-2-oxide
461350010
461350050
ACRO461350010EA
142
EUR
ACRO461350010
ACRO461350050
Ifosfamide
Ifosfamide
Formel:
C₇H₁₅Cl₂N₂O₂P Molecular Weight: 261,09 g/mol Schmelzpunkt: 39…42 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00057374 CAS-Nummer: 3778-73-2 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
HPLC | ≥97.5 % (ELSD) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...