Desvenlafaxine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(2-(Dimethylamino)-1-(1-hydroxycyclohexyl)-ethyl)phenol, O-Desmethylvenlafaxine, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, Desvenlafaxine

461570010 461570050
ACRO461570010EA 51.3 EUR
ACRO461570010 ACRO461570050
Desvenlafaxine 97%
Desvenlafaxine
Formel: C₁₆H₂₅NO₂
Molecular Weight: 263,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00871934
CAS-Nummer: 93413-62-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR