Caffeine citrate
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 2-hydroxypropane-1,2,3-tricarboxylic acid, citric acid,1,3,7-trimethylpurine-2,6-dione, Citrated caffeine
461720250 461721000
ACRO461720250EA 105 EUR
ACRO461720250 ACRO461721000
Caffeine citrate
Caffeine citrate
Formel: C₁₄H₁₈N₄O₉
Molecular Weight: 386.32 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00044986
CAS-Nummer: 69-22-7
Bestellen
Spezifikation Testergebnisse
Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Titration with HClO4 48 to 52 % (Caffeine)
Titration with TBAH 48 to 52 % (Citric acid)