Clofazimin 98%
Lieferant: Thermo Scientific
Synonyme:
N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine, N,5-Bis(4-chlorophenyl)-3-(isopropylimino)-3,5-dihydrophenazin-2-amine
461760010
461760050
ACRO461760010EA
61.3
EUR
ACRO461760010
ACRO461760050
Clofazimin 98%
Clofazimine
Formel:
C₂₇H₂₂Cl₂N₄ Molecular Weight: 473,40 g/mol Schmelzpunkt: 219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056793 CAS-Nummer: 2030-63-9 |
Spezifikation Testergebnisse
Appearance | Orange to red to brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C) |
HPLC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...