Triclabendazole 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)-1H-benzo[d]imidazole, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole, 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole
461960010
461960050
ACRO461960010EA
72.8
EUR
ACRO461960010
ACRO461960050
Triclabendazole 98%
Triclabendazole
Formel:
C₁₄H₉Cl₃N₂OS Molecular Weight: 359.66 g/mol Schmelzpunkt: 175…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00864519 CAS-Nummer: 68786-66-3 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystallinje powder or crystals |
Infrared spectrum | Conforms |
Melting point | 175 to 180 °C |
Loss on drying | ≤1 % (1 g, 105 °C, 6 h) |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...