Emtricitabin

Lieferant: Thermo Scientific

Synonyme: 4-Amino-5-fluoro-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one

462070010 462070050
ACRO462070010EA 87.1 EUR
ACRO462070010 ACRO462070050
Emtricitabin
Emtricitabine
Formel: C₈H₁₀FN₃O₃S
Molecular Weight: 247,24 g/mol
Schmelzpunkt: 136…140 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00870151
CAS-Nummer: 143491-57-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 153 to 157 °C
Loss on drying ≤0.5 %
HPLC ≥96.0 %
Specific optical rotation -125° to -142° (20 °C, 589 nm) (0.25 % in Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR