Levetiracetam

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-2-(2-Oxopyrrolidin-1-yl)butanamide, (S)-2-(2-Oxo-1-pyrrolidinyl)butyramide

462120050 462120250
ACRO462120050EA 69.3 EUR
ACRO462120050 ACRO462120250
Levetiracetam
Levetiracetam
Formel: C₈H₁₄N₂O₂
Molecular Weight: 170,21 g/mol
Schmelzpunkt: 117 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03265610
CAS-Nummer: 102767-28-2

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 116 to 120 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation -76° to -82° (20 °C, 589 nm) (2 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR