Triamcinolone

Lieferant: Thermo Scientific
Achtung

Synonyme: 9alpha-Fluoro-11beta,16alpha,17,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, Fluoxyprednisolone

462240010 462240050
ACRO462240010EA 98.5 EUR
ACRO462240010 ACRO462240050
Triamcinolone
Triamcinolone
Formel: C₂₁H₂₇FO₆
Molecular Weight: 394,44 g/mol
Schmelzpunkt: 262…263 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 124-94-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR