BQCA ≥98%, kristallin

Lieferant: Cayman Chemical

Synonyme: 1-(4-Methoxybenzyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, Benzyl quinolone carboxylic acid.

15393-10 15393-5 15393-25 15393-50
CAYM15393-10EA 128 EUR
CAYM15393-10 CAYM15393-5 CAYM15393-25 CAYM15393-50
BQCA ≥98%, kristallin
BQCA

BQCA is a highly selective positive allosteric modulator of the M₁ muscarinic acetylcholine receptor (mAChR), as it dose-dependently reduces the concentration of acetylcholine required to activate the M₁ receptor. The effective range for potentiation of M₁ in cells by BQCA is 0,1 to 100 μM with an inflection point value of 845 nM when 3 nM acetylcholine is used.


BQCA displays no potentiation, agonism, or antagonism at other mAChRs at concentrations up to 100 μM. It has excellent brain penetration and increases the firing rate of medial prefrontal cortex neurons in vivo in rats. BQCA prevents scopolamine-induced memory deficits in both contextual fear conditioning and a spontaneous alternation task in mice. It also restores impairment in reversal learning in a mouse model of Alzheimer’s disease and improves memory performance in rats.

Formel: C₁₈H₁₅NO₄
Molecular Weight: 309.3 g/mol
MDL: MFCD01315710
CAS-Nummer: 338747-41-4

Bestellen

Spezifikation Testergebnisse

Purity ≥98%
Formulation A crystalline solid
λmax 224, 315, 328 nm
Storage –20 °C
Stability ≥2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR