4-Methoxyzimtsäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(4-Methoxyphenyl)acrylic acid
H60144.14
H60144.22
H60144.14EA
59.5
EUR
H60144.14
H60144.22
4-Methoxyzimtsäure ≥99%
4-Methoxyzimtsäure
Purity: 99%
CAS No.: 830-09-1
Molecular Formula: C10H10O3
Formula Weight: 178.18
Melting Point: 170-173°
MDL No.: MFCD00004398
Formel:
C₁₀H₁₀O₃ Molecular Weight: 178,18 g/mol Schmelzpunkt: 170…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004398 CAS-Nummer: 830-09-1 EINECS: 212-594-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...