Verapamil hydrochloride ≥98%
Lieferant: Cayman Chemical
Synonyme: 5-((3,4-Dimethoxyphenethyl)(methyl)amino)-2-(3,4-dimethoxyphenyl)-2-isopropylpentanenitrile hydrochloride, Verapamil HCl, 5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile Hydrochloride, (±)-Verapamil hydrochloride
14288-1 14288-5
CAYM14288-1EA 39 EUR
CAYM14288-1 CAYM14288-5
Verapamil hydrochloride ≥98%
Verapamil hydrochloride
Verapamil is the prototypical blocker of L-type calcium channels that produces excitation-contraction uncoupling in cardiac muscle by preventing the slow-inward current of calcium ions.
Verapamil can also block calcium fluxes in vascular smooth muscle. It has both peripheral and coronary vasodilator effects (IC50=0,38 μm in guinea pig aortic strip) and has been used to control hypertension, angina, cardiac arrhythmia, and vascular headaches. Verapamil has also been used in cell biology as an inhibitor of drug efflux pump proteins such as P-glycoprotein, which are often over-expressed in certain tumor cell lines.
Formel: C₂₇H₃₈N₂O₄
Molecular Weight: 491.07 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00055208
CAS-Nummer: 152-11-4