(4-Nitrophenyl)-β-D-galactopyranosid ≥98%

Lieferant: Abcam

Synonyme: pNPG, 4-Nitrophenyl β-D-Galactopyranoside

AB146393-1 AB146393-5
ABCAAB146393-1EA 145 EUR
ABCAAB146393-1 ABCAAB146393-5
(4-Nitrophenyl)-β-D-galactopyranosid ≥98%
(4-Nitrophenyl)-β-D-galactopyranosid

Chromogenic β-galactosidase substrate yielding a yellow end product upon cleavage.


  • Source: synthetic
  • Store at −20 °C


Synonyms: 4-Nitrophenyl-β-D-galactopyranoside; 4-Nitrophenyl galactoside; p-Nitrophenyl-beta-D-galactoside

Formel: C₁₂H₁₅NO₈
Molecular Weight: 301.25 g/mol
MDL: MFCD00063256
CAS-Nummer: 3150-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR