(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride, Sapropterin 2HCl, Biopten, Sapropterin dihydrochloride, (R)-2-Amino-6-((1R,2S)-1,2-dihydroxypropyl)-5,6,7,8-tetrahydropteridin-4(1H)-one dihydrochloride
328871000
328875000
ACRO328871000EA
171
EUR
ACRO328871000
ACRO328875000
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
Formel:
C₉H₁₅N₅O₃·2HCl Molecular Weight: 314,17 g/mol Schmelzpunkt: 250…255 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00891665 CAS-Nummer: 69056-38-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...