Chlorothiazid 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-Chloro-2H-1,2,4-benzothiadiazine-7-sulphonamide 1,1-Dioxide
461740050
461740250
461741000
ACRO461740050EA
37.9
EUR
ACRO461740050
ACRO461740250
ACRO461741000
Chlorothiazid 98%
Chlorothiazide
Formel:
C₇H₆ClN₃O₄S₂ Molecular Weight: 295,71 g/mol Schmelzpunkt: 348 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00058576 CAS-Nummer: 58-94-6 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...