Isoprenaline hemisulphate
Lieferant: Thermo Scientific
Synonyme:
4-[1-Hydroxy-2-[(1-isopropyl)amino]ethyl]-1,2-benzenediol hemisulphate
462110050
462110250
ACRO462110050EA
43.8
EUR
ACRO462110050
ACRO462110250
Isoprenaline hemisulphate
Isoprenaline hemisulphate
Formel:
C₁₁H₁₇NO₃·¹/₂H₂O₄S Molecular Weight: 260,30 g/mol Schmelzpunkt: 83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013139 CAS-Nummer: 299-95-6 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Water | ≤7 % |
HPLC | ≥97.5 % |
Identity | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...