Losartan-Kalium (durch HPLC)
Lieferant: Thermo Scientific
Synonyme:
MK-954, DUP-753, 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)-1,1'-biphenyl-4-yl]methyl]imidazole-5-methanol Potassium Salt, (1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-4-chloro-1H-imidazol-5-yl)methanol, potassium salt
462740050
462740250
ACRO462740050EA
84.9
EUR
ACRO462740050
ACRO462740250
Losartan-Kalium (durch HPLC)
Losartan-Kalium
Formel:
C₂₂H₂₃ClN₆O Molecular Weight: 461,01 g/mol Schmelzpunkt: 183…185 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02092704 CAS-Nummer: 124750-99-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C) |
Titration with HClO4 | 98.5 to 101.5 % (on dried substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...