Saxagliptin
Lieferant: Thermo Fisher Scientific
Synonyme:
(1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile
462180010
462182500
ACRO462180010EA
394
EUR
ACRO462180010
ACRO462182500
Saxagliptin
Saxagliptin
Formel:
C₁₈H₂₅N₃O₂ Molecular Weight: 315.42 g/mol Schmelzpunkt: 107…110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12756398 CAS-Nummer: 361442-04-8 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...