Vindoline

Lieferant: Thermo Scientific
Achtung

462270010 462270050
ACRO462270010EA 87.4 EUR
ACRO462270010 ACRO462270050
Vindoline
Vindoline
Formel: C₂₅H₃₂N₂O₆
Molecular Weight: 456,5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD27991347
CAS-Nummer: 2182-14-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR