DL-Mevalonsäurelacton ∼97% (durch titrimetrische Analyse), Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-Mevalonolactone, 4-Hydroxy-4-methyltetrahydro-2H-pyran-2-one, β-Hydroxy-beta-methyl-delta-valerolactone, DL-Mevalonic Acid Lactone, Tetrahydro-4-hydroxy-4-methyl-2-pyrone

M-Clarity™ Qualitätsstufe = MQ200

M4667-10G M4667-1G M4667-5G
SIALM4667-10GEA 1210 EUR
SIALM4667-10G SIALM4667-1G SIALM4667-5G
DL-Mevalonsäurelacton ∼97% (durch titrimetrische Analyse), Sigma-Aldrich®
DL-Mevalonolactone

Used to study the effects of statin on the prenylation of Ras and Rho GTPases, statins on proliferation and migration of HUVECs (HGF-induced human umbilical vein endothelial cells) and to analyse the isoprenoid biosynthesis pathways in Listeria monocytogenes.

Formel: C₆H₁₀O₃
Molecular Weight: 130,14 g/mol
MDL: MFCD00006648
CAS-Nummer: 674-26-0

Bestellen

Spezifikation Testergebnisse

Assay (titration) 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR