SLPI (human), ELISA kit
Lieferant: Enzo Life Sciences
ENZOADI9002220001EA 866.8 EUR
ENZOADI9002220001 ALEXADI9002220001
SLPI (human), ELISA kit
Tests ELISAs
Rapid ELISA kit to quantify SLPI, a secretory leukocyte peptidase inhibitor and unique cancer biomarker.
For the quantitative determination of human SLPI (ALP, Antileukoproteinase) in serum.
The SLPI ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of SLPI in serum with results in just 2.5 hours.
Bestellinformation: Contains Microtiter Plate, Standards, Calibrators, Antibody-conjugate, TMB Reagent, Stop Solution, Wash Buffer Concentrate.