ETA receptor antagonist [BQ-610]
Lieferant: Enzo Life Sciences
ALEXALX155011P001EA 193 EUR
ALEXALX155011P001 ENZOALX155011P001
ETA receptor antagonist [BQ-610]
Proteine und Peptide
30-fold more potent ETA antagonist than BQ-123 in pig aortic smooth muscle membrane assay, same activity as BQ-123 when tested on pig cerebellum membrane.
Bestellinformation: Quantity: *Sold as net peptide weight.