Supplements for renal epithelial cell growth medium 2
Lieferant: PROMOCELL
C-39606
C-39605
C-39606EA
164
EUR
C-39606
C-39605
Supplements for renal epithelial cell growth medium 2
Zellkultur-Ergänzungen und -Zusätze
PromoCell Renal Epithelial Cell Growth Medium 2 is an optimised low serum medium that contains epinephrine. The medium consists of Basal Medium plus SupplementMix. Supplements are also available to purchase separately, either in separate vials (SupplementPack) or as a pre-prepared mix (SupplementMix).