Diiodo(p-cymene)ruthenium(II) dimer, gelb, kristallin
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Di-µ-iodobis[(p-cymene)iodoruthenium(II)]
46886.03
46886.06
46886.03EA
110
EUR
46886.03
46886.06
Diiodo(p-cymene)ruthenium(II) dimer, gelb, kristallin
Diiodo(p-cymene)ruthenium(II) dimer
- Sensitivity: Moisture and light sensitive
- Storage: Store under nitrogen
Diiodo(p-cymene)ruthenium(II) dimer can be applied as a catalyst for many reactions such as ruthenium-catalysed hydrogenation, preparation of Walphos-type ferrocenyl-aryl diphosphine ligands, ruthenium-catalysed synthesis reactions, and enantioselective hydrogenation of ketones and olefins.
Insoluble in water.
Formel:
C₂₀H₂₈I₄Ru₂ Molecular Weight: 978.20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08064027 CAS-Nummer: 90614-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...