Natriumperiodat 0.1 mol/l in Wasser Reag. Ph. Eur.

Lieferant: Bernd Kraft
Gefahr

Synonyme: Natriumtetroxoiodat(VII), Natriummetaperiodat

25042.3100 25042.4100
KRAF25042.3100EA 214 EUR
KRAF25042.3100 KRAF25042.4100
Natriumperiodat 0.1 mol/l in Wasser Reag. Ph. Eur.
Natriumperiodat
Formel: NaIO₄
Molecular Weight: 213,89 g/mol
MDL: MFCD00003534
CAS-Nummer: 7790-28-5
EINECS: 232-197-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR