Magnesium ≥99,8% (Metall-Basis), Drehspäne

Lieferant: Thermo Fisher Scientific
Gefahr

10235.22 10235.36
10235.22EA 57 EUR
10235.22 10235.36
Magnesium ≥99,8% (Metall-Basis), Drehspäne
Magnesium

Purity: 99.8% Physical Form: 2.3mm (0.09in) wide Appearance: Silver-grey Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308

Formel: Mg
Molecular Weight: 24.3 g/mol
Siedepunkt: 1107
Schmelzpunkt: 651
Lagertemperatur: Raumtemperatur
MDL: MFCD00085308
CAS-Nummer: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR