Aluminium ≥99,998% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)
Lieferant: Thermo Scientific
Synonyme:
Aluminum
11373.FI
11373.GH
11373.HP
11373.FIEA
73.6
EUR
11373.FI
11373.GH
11373.HP
Aluminium ≥99,998% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)
Aluminium
Purity: 99.998%
CAS No.: 7429-90-5
Physical Form: ≈6.75g/50x50mm, Temper: as rolled
Odour: Odourless
MDL No.: MFCD00134029
Formel:
Al Molecular Weight: 26,98 g/mol Siedepunkt: 2327 °C (1013 hPa) Schmelzpunkt: 660 °C Dichte: 2,7 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134029 CAS-Nummer: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...