Aluminium ≥99,999% (Metall-Basis), Stäbe, Puratronic®, Ø 6.35 mm (0.25 in)

Lieferant: Thermo Fisher Scientific

11493.BQ 11493.BU 11493.CZ
11493.BQEA 104 EUR
11493.BQ 11493.BU 11493.CZ
Aluminium ≥99,999% (Metall-Basis), Stäbe, Puratronic®, Ø 6.35 mm (0.25 in)
Aluminium

Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈0.87g/cm Odour: Odourless MDL No.: MFCD00134029

Formel: Al
Siedepunkt: 2327 °C (1013 hPa)
Schmelzpunkt: 660 °C
Dichte: 2,7 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134029
CAS-Nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR