Gallium 99,9999 Metals basis, ingot

Lieferant: Thermo Fisher Scientific
Achtung

10185.18 10185.14
10185.18EA 362 EUR
10185.18 10185.14
Gallium 99,9999 Metals basis, ingot
Gallium

Purity: 99.9999% CAS No.: 7440-55-3 Storage Temperature: 2°C to 8°C Appearance: Silver grey UN No.: 2803 MDL No.: MFCD00134045

Formel: Ga
Molecular Weight: 69.72 g/mol
Siedepunkt: 2403
Schmelzpunkt: 29
Dichte: 5.904 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00134045
CAS-Nummer: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8 III
Merck Index: 12,04364

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR