Indium ≥99,999% (Metall-Basis), Pulver, Puratronic® -325 mesh
Lieferant: Thermo Fisher Scientific
Achtung
00151.06 00151.14
00151.06EA 146 EUR
00151.06 00151.14
Indium ≥99,999% (Metall-Basis), Pulver, Puratronic® -325 mesh
Indium
Purity: 99.999% CAS No.: 7440-74-6 Molecular Formula: In Formula Weight: 114.82 Physical Form: Powder Density: 7.31 Appearance: Silver-grey Odour: Odourless Melting Point: 156.6° Boiling Point: 2072° UN No.: 3089 MDL No.: MFCD00134048
Formel: In
Molecular Weight: 114.82 g/mol
Siedepunkt: 2072 °C
Schmelzpunkt: 156.6 °C
Dichte: 7.31
Lagertemperatur: Raumtemperatur
MDL: MFCD00134048
CAS-Nummer: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4.1 III
Merck Index: 13,04971