Germanium ≥99,999% (Metall-Basis), Pulver -100 mesh
Lieferant: Thermo Scientific
10190.09
10190.18
10190.09EA
233
EUR
10190.09
10190.18
Germanium ≥99,999% (Metall-Basis), Pulver -100 mesh
Germanium
Purity: 99.999%
CAS No.: 7440-56-4
Odour: Odourless
MDL No.: MFCD00085310
Formel:
Ge Molecular Weight: 72,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085310 CAS-Nummer: 7440-56-4 EINECS: 231-164-3 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...