Germanium ≥99,9999% (Metall-Basis), Stücke, Puratronic® ≤ 2 cm (0.8 in)

Lieferant: Thermo Scientific

11144.06 11144.14 11144.22
11144.06EA 129 EUR
11144.06 11144.14 11144.22
Germanium ≥99,9999% (Metall-Basis), Stücke, Puratronic® ≤ 2 cm (0.8 in)
Germanium

Purity: 99.9999+% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odour: Odourless MDL No.: MFCD00085310

Formel: Ge
Molecular Weight: 72,61 g/mol
Siedepunkt: 2830 °C (1013 hPa)
Schmelzpunkt: 937,2 °C
Dichte: 5,323 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00085310
CAS-Nummer: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR