Arsen ≥99,99999% (Metall-Basis), polykristalliner Brocken, Puratronic® 2-8 mm (0.08-0.3 in)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Arsenic solution
unter Argon abgefüllt
13044.03
13044.09
13044.14
13044.22
13044.A1
13044.03EA
33.6
EUR
13044.03
13044.09
13044.14
13044.22
13044.A1
Arsen ≥99,99999% (Metall-Basis), polykristalliner Brocken, Puratronic® 2-8 mm (0.08-0.3 in)
Arsenic
Purity: 99.99999+%
CAS No.: 7440-38-2
Molecular Formula: As
Formula Weight: 74.92
Physical Form: Packaged under argon
Sensitivity: Air Sensitive
Appearance: Grey
Odour: Odourless
UN No.: 1558
MDL No.: MFCD00085309
Formel:
As Molecular Weight: 74.92 g/mol Schmelzpunkt: 817 Dichte: 5.727 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085309 CAS-Nummer: 7440-38-2 EINECS: 213-148-6 UN: 1558 ADR: 6.1 II Merck Index: 12,00832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...