Europium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Barren

Lieferant: Thermo Scientific
Gefahr

00120.03 00120.06
00120.03EA 161 EUR
00120.03 00120.06
Europium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Barren
Europium

Purity: 99.9% CAS No.: 7440-53-1 Physical Form: Packaged under mineral oil Appearance: Grey Odour: Odourless UN No.: 3208 MDL No.: MFCD00010992

Formel: Eu
Molecular Weight: 151,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010992
CAS-Nummer: 7440-53-1
EINECS: 231-161-7
UN: 3208
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR