Holmium ≥99,9% (Metall-Basis ausgeschlossen Ta), Stücke
Lieferant: Thermo Scientific
00142.06
00142.14
00142.06EA
180
EUR
00142.06
00142.14
Holmium ≥99,9% (Metall-Basis ausgeschlossen Ta), Stücke
Holmium
Purity: 99.9%
CAS No.: 7440-60-0
Physical Form: Packaged in air-stable poly bottle
MDL No.: MFCD00011049
Formel:
Ho Molecular Weight: 164,93 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011049 CAS-Nummer: 7440-60-0 EINECS: 231-169-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...