Holmium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.127 mm (0.005 in)
Lieferant: Thermo Scientific
00144.FF
00144.FI
00144.FFEA
306
EUR
00144.FF
00144.FI
Holmium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.127 mm (0.005 in)
Holmium
Purity: 99.9%
CAS No.: 7440-60-0
Physical Form: ≈0.70g/25x25mm, Packaged in mylar under argon
Odour: Odourless
MDL No.: MFCD00011049
Formel:
Ho Molecular Weight: 164,93 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011049 CAS-Nummer: 7440-60-0 EINECS: 231-169-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...