Holmium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.3 mm (0.01 in)

Lieferant: Thermo Fisher Scientific

10210.FF 10210.FI
10210.FFEA 331 EUR
10210.FF 10210.FI
Holmium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.3 mm (0.01 in)
Holmium

Purity: 99.9% CAS No.: 7440-60-0 Physical Form: ≈1.65g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011049

Formel: Ho
Molecular Weight: 164.93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011049
CAS-Nummer: 7440-60-0
EINECS: 231-169-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR