Holmium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.025 mm (0.001 in)

Lieferant: Thermo Scientific
Gefahr

42687.FF 42687.FI
42687.FFEA 300 EUR
42687.FF 42687.FI
Holmium ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.025 mm (0.001 in)
Holmium

Purity: 99.9% CAS No.: 7440-60-0 Physical Form: ≈0.14g/25x25mm, Packaged in mylar under argon Appearance: Gray to silver Odour: Odourless UN No.: 3178 MDL No.: MFCD00011049

Formel: Ho
Molecular Weight: 164,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011049
CAS-Nummer: 7440-60-0
EINECS: 231-169-0
UN: 3178
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR