Holmium ≥99,9% (Metall-Basis), Stäbe, Ø 6.35 mm (0.25 in)
Lieferant: Thermo Fisher Scientific
44050.DJ
44050.HF
44050.DJEA
1056
EUR
44050.DJ
44050.HF
Holmium ≥99,9% (Metall-Basis), Stäbe, Ø 6.35 mm (0.25 in)
Holmium
Purity: 99.9%
CAS No.: 7440-60-0
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.28g/mm, Cast rod, diameter is approximate
Odour: Odourless
Formel:
Ho Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011049 CAS-Nummer: 7440-60-0 EINECS: 231-169-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...