Scandium, Pulver, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

261262-1G 261262-250MG
SIAL261262-1GEA 784 EUR
SIAL261262-1G SIAL261262-250MG
Scandium, Pulver, Sigma-Aldrich®
Scandium
Formel: Sc
Molecular Weight: 44,96 g/mol
MDL: MFCD00016323
CAS-Nummer: 7440-20-2
EINECS: 231-129-2
UN: 3178
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR