Tantal ≥99,9% (Metall-Basis), Drahtform, Ø 0.127 mm (0.005 in)

Lieferant: Thermo Scientific

00977.G1 00977.G5
00977.G1EA 63.2 EUR
00977.G1 00977.G5
Tantal ≥99,9% (Metall-Basis), Drahtform, Ø 0.127 mm (0.005 in)
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈0.21g/m Odour: Odourless MDL No.: MFCD00011252

Formel: Ta
Molecular Weight: 180,95 g/mol
Siedepunkt: 5427 °C (1013 hPa)
Schmelzpunkt: 2996 °C
Dichte: 16,69 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011252
CAS-Nummer: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR