Tantal hart ≥99,95% (Metall-Basis), Folie, hart, Dicke 0.025 mm (0.001 in)

Lieferant: Thermo Scientific

10351.IV 10351.IX 10351.GJ
10351.IVEA 475 EUR
10351.IV 10351.IX 10351.GJ
Tantal hart ≥99,95% (Metall-Basis), Folie, hart, Dicke 0.025 mm (0.001 in)
Tantalum

Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈6.3g/100x150mm, Temper: as rolled Odour: Odourless MDL No.: MFCD00011252

Formel: Ta
Molecular Weight: 180,95 g/mol
Siedepunkt: 5427 °C (1013 hPa)
Schmelzpunkt: 2996 °C
Dichte: 16,69 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011252
CAS-Nummer: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR